Learn More
Fisherbrand™ SPP Disposable Beard Cover
General purpose, high quality, non-woven disposable beard covers that are made of spunbound polypropylene material, ideal for use in the laboratory or elsewhere for promoting general hygiene.
Brand:  Fisherbrand™ FBHMBC
55.86 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Product Code. 17273034
  • 81.12 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
General purpose beard covers with integral elasticated headloop and seam for a comfortable, universal fit.
  • Non-woven, lightweight spunbound polypropylene at 12 g/m2
  • For use in the laboratory or wider working environment
  • Manufactured and vacuum-sealed in a stringent clean environment
  • Latex-free
  • ISO tested to ISO3801 and ISO13934-1
Specifications
Specifications
Beard Cover
Spunbound Polypropylene
Yes
Elastic
General Purpose in the Lab or Cleanroom
23 ±2 cm; 46 ±2 cm Stretched
12 gsm
Large/Universal
White
Yes
PPE CAT I, ISO Tested To ISO3801 and ISO13934-1; Meets 4.0 AQL Level II Sampling Plan Per MIL-std-150E For Defect Criteria
1000 Pk.
10 cm ±2 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ SPP Disposable Beard Cover > SPP Disposable Beard Cover; Certifications/Compliance: PPE CAT I, ISO Tested To ISO3801 and ISO13934-1; Length (Metric): 23 ±2 cm; 46 ±2 cm Stretched; Material: Spunbond Polypropylene