Learn More
Edmund Bühler GmbH™ Spring Clamp
Fasten Erlenmeyer flasks on the universal tray.
Brand:  Edmund Bühler GmbH™ 0009 645
 View more versions of this product
Product Code. 10757441
  • 23.30 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel construction
  • For fastening Erlenmeyer flasks on the universal tray
  • The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
Specifications
100 mL
Spring Clamp
Stainless Steel