missing translation for 'onlineSavingsMsg'
Learn More

Julabo™ Spring Clamps for Shaking Water Bath

Brand:  Julabo™ 8970631

 View more versions of this product

Product Code. 10833961

  • 88.95 € / Each
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Twelve 135 mm and five 190mm L springs
Basic tray 8970630, SW22, SW23 Shaking water bath
Spring Clamp Set
12 Spring clamps 135mm and 5 spring clamps 190mm L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers