Learn More
DWK Life Sciences Square Quadrupod, for glass ceramic laboratory protection plate
Ideal holder for glass ceramic plates.
Brand:  DWK Life Sciences 290775302
235.93 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15262109
  • 260.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • holder plate: steel 1.4301, tripod feet: steel 1.4305
  • Made from heat-resistant chrome-nickel steel with
  • Four legs for extra stability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chrome-nickel Steel
Plate holder for glass ceramic plates; Heating; For Glass Ceramic Laboratory Protection Plate
5/Pk.
135 mm
210 mm