missing translation for 'onlineSavingsMsg'
Learn More
SR 16584, Tocris Bioscience™ Product Code.: 30001475

SR 16584, Tocris Bioscience™

Product Code. 30001475
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001475

Brand: Tocris Bioscience 4424/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective α3β4 nAChR antagonist

Selective α3β4 nAChR antagonist (IC50 = 10.2 μM). Selectively binds α3β4 over α4β2 and α7 subtypes (Ki values are 0.508, >100 and >100 μM, respectively).

Chemical Identifiers

CAS 1150153-86-8
Molecular Formula C17H22N2O
Molecular Weight (g/mol) 270.376
InChI Key GIDZCNCCCWFCIN-YIONKMFJSA-N
Synonym 1,3-dihydro-1-3-exo-9-methyl-9-azabicyclo 3.3.1 non-3-yl-2h-indol-2-one, 1-1r,3s,5s-9-methyl-9-azabicyclo 3.3.1 nonan-3-yl-3h-indol-2-one
PubChem CID 49840264
IUPAC Name 1-[(1S,5R)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]-3H-indol-2-one
SMILES CN1C2CCCC1CC(C2)N3C(=O)CC4=CC=CC=C43

Specifications

Quantity 10 mg
Formula Weight 270.37
Percent Purity >98%
Chemical Name or Material SR 16584
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SR 16584, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.