missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 9243
Selective LXR inverse agonist
201.00 € - 749.00 €
Specifications
Color | White |
---|---|
Product Type | Inhibitor |
CAS | 1613028-81-1 |
Molecular Formula | C31H32BrNO4S2 |
Formula Weight | 626.62g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797536
|
Tocris Bioscience™
6138/10 |
10 mg |
201.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746596
|
Tocris Bioscience™
6138/50 |
50 mg |
749.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective LXR inverse agonist. Exhibits no significant activity at other nuclear receptors at maximally effective concentration. Inhibits the Warburg effect and lipogenesis by down-regulation of LXR-mediated gene expression. Selectively reduces cell viability and induces apoptosis in cancer cell lines in vitro. Suppresses hepatic steatosis and inhibits growth of tumor xenografts in mice.Specifications
White | |
1613028-81-1 | |
626.62g/mol | |
Store at 4°C |
Inhibitor | |
C31H32BrNO4S2 | |
Soluble to 20mM in DMSO | |
Inhibits IL-1β |
For Research Use Only