Learn More
SRGAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22519-1-AP-20UL
 View more versions of this product
Product Code. 16811965
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
Specifications
Specifications
SRGAP2
Polyclonal
Unconjugated
SRGAP2
ARHGAP34, FNBP2, Formin binding protein 2, KIAA0456, SRGAP2, srGAP3
Rabbit
Antigen Affinity Chromatography
RUO
14270, 23380
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75044, Q91Z67
SRGAP2
SRGAP2 Fusion Protein Ag18278
20 μL
Primary
Mouse, Human