missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP9 Mouse anti-Human, Mouse, Rat, Clone: 1H7G10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66068-1-IG-20UL
This item is not returnable.
View return policy
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.Specifications
SRP9 | |
Monoclonal | |
0.93 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49458, P49962 | |
SRP9 | |
SRP9 Fusion Protein Ag17153 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Western Blot | |
1H7G10 | |
Unconjugated | |
SRP9 | |
ALURBP, SRP9 | |
Mouse | |
Protein A | |
RUO | |
27058, 6726, 690345 | |
-20°C | |
Liquid |