missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5112725
Additional Details : Weight : 0.01000kg
Description
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.Specifications
SRR | |
Polyclonal | |
Unconjugated | |
Srr | |
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
Rabbit | |
Protein G | |
RUO | |
27364, 63826 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9GZT4, Q9QZX7 | |
Srr | |
Recombinant Human Serine racemase protein (1-340AA) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |