Learn More
SRRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12822-1-AP-20UL
 View more versions of this product
Product Code. 16810744
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human SRRM1.
Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3′-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.
Specifications
Specifications
SRRM1
Polyclonal
Unconjugated
SRRM1
160 KD, POP101, SRM160, SRRM1
Rabbit
Antigen Affinity Chromatography
RUO
10250, 313620, 51796
-20°C
Liquid
Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q52KI8, Q8IYB3
Srrm1
SRRM1 Fusion Protein Ag3513
20 μL
Primary
Mouse, Human, Rat