missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SSC buffer (20X)

Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

47.74 € - 133.67 €

Specifications

Content And Storage Ambient temperatures
Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413549
View Documents
Thermo Scientific Chemicals
J60839.K2
1 L
47.74 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15433549
View Documents
Thermo Scientific Chemicals
J60839.K7
4 L
133.67 €
4 L
Please to purchase this item. Need a web account? Register with us today!
15423549
View Documents
Thermo Scientific Chemicals
J60839.K3
2 L
73.67 €
2 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ambient temperatures
Liquid
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.