missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ ST-20 Tube
Can be used for mixing, stirring, extractions, preparation of soil sample suspensions and solubility testing for medication.
Brand: IKA™ 0003703000
This item is not returnable.
View return policy
Description
Contains: tube, stirring device and cover
Specifications
ST-20 Tube | |
Mixing, stirring, extractions, preparation of soil sample suspensions, solubility testing for medication | |
1 Ea. |
2 to 15 mL | |
ULTRA-TURRAX™ Tube Drive |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKA™ ST-20 Tube > Capacity: 2 to 15mL, Quantity: 1Each
Spot an opportunity for improvement?Share a Content Correction