Learn More
Kinematica™ ST-P20/600 Plate Stand for Polytron™ PT 10-35 GT Homogenizers
Plate stand with two support rods. The drive unit‘s height can be adjusted. For more security, the drive unit cannot be adjusted horizontally.
Brand:  Kinematica™ 11040051
Product Code. 12719336
  • 477.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Plate Stand
Polytron PT 10-35 GT Homogenizer
ST-P20/600