Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
54.15 € - 74.25 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS Compliant to 2011/65/EU (Pass 2015/863)
Adhesive Type Permanent Acrylic
Coating None
Color Black on White
View More Specs
Products 4
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
17821789
View Documents
Brady™
175660
0.25 x 1.05 in.
74.25 €
Pack of 360
17871719
View Documents
Brady™
170885
0.375 x 1 in.
54.50 €
Pack of 320
17811799
View Documents
Brady™
170886
0.75 x 0.9 in.
58.20 €
Pack of 320
17851759
View Documents
Brady™
170884
0.9 x 0.9 in.
54.15 €
Pack of 280
Description
Description
  • Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
  • Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Specifications
Specifications
Autoclavable
Permanent Acrylic
Black on White
Die-cut
Top
Labels
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling
-112°F to +266°F
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
None
Matte
Polyester
Thermal Transfer
Non-sterile
BMP51, M511
0.0033 in.