Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Laboratory Labels for M6/M7 Printers
Super chemical-resistant polyester is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
Brand:  Brady™ 174088
 View more versions of this product
Product Code. 17838284
  • 68.00 € / Pack of 250
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process
  • High resistance to a wide variety of chemicals, oils and water
  • Service temperature of -80°C to +130°C (-112°F to +266°F)
  • Made of B-481 polyester material
Specifications
Specifications
Autoclavable
M6-194-481
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
250 Labels
-112°F to +266°F
0.0033 in.
0.75 x 0.9 in.
0.9 in.
19.05 mm
ASTM D 1000, ASTM D 2979
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling
No
Rectangle
-80°C to +130°C
0.084 mm
0.75 in.
22.86 mm