Learn More
Kartell™ Staining Jar with 2 Lids
Kartell™ Staining Jar with 2 Lids provides evaporation free storage of staining liquids and allows a staining rack handle to be inserted.
Brand:  Kartell™ 0035300
Product Code. 10419412
  • 122.00 € / Pack of 4
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded in PMP
  • Supplied with 2 lids
  • One lid for use when dish is not in use, providing evaporation free storage of staining liquid
  • The other lid with special opening to allow handle of staining rack to be inserted
  • Xylene can cause etching after 2 days at room temperture
Specifications
Specifications
Closure Included
Flat
PMP
4/Pk.
74 mm
63 mm
PMP
Clear
Staining Jar
95 x 74 mm
95 mm