missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Staining Trough

Allows for the staining of 10 microscope slides.

Brand:  Karl Hecht™ 41205002

 View more versions of this product

Product Code. 10573452

  • 21.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Glass trough with cover
  • Staining tray made of stainless steel
  • Suitable for microscope slides of approx. 76 x 26mm, width up to approx. 45mm
  • Spare part of glass trough also available
Specifications

Specifications

Spare trough
Staining slides
9 x 7 x 6.5 cm
Clear
Slides of approx. 76 x 26 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.