Learn More
BRAND™ Staining Troughs with Tray
For 25 slides
Brand:  BRAND™ 471800
Product Code. 10242191
Description
Description
  • The close fitting lid helps protect against evaporation
  • The staining tray has a fold down handle
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Universal Slide Staining Kit
    25
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    BRAND™ Staining Troughs with Tray > Dimensions (L x W): 98 x 88 mm