missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 3 Model 100W Cooled Incubator, 100 L, Stainless Steel

Temperature Range: -10°C to +50°C

Brand:  LMS™ 100

Product Code. 11872063

  • 5865.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
  • Reliable and accurate temperature control
  • Standard single and dual temperature, plus programmable models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications

Specifications

ISO 9001:2008
Freestanding incubator
Stainless Steel
500 mm x 380 mm
Stove Enameled Mild Steel
-10°C to +50°C
Digital
100W
CFC-free
±0.1°C
2 Years
21.3 in.
440 mm
19.3 in.
490 mm
440 mm
240 V, 50 Hz
50 Hz
Standard digital
100 L
Fan assisted air circulation via premixing chamber
Optional Fluorescent/UV
2
Refrigerated Incubator
Foamed polyurethane
1
500 x 380 mm
±0.5°C
24.0 in.
540 mm
32.3 in.
820 mm
17.3 in.
50 kg
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers