Learn More
Usbeck Laborgeraete™ Stainless Steel Alcohol Burners
Product Code. p-8008735
Click to view available options
Includes:
Adjustable Wick and Cap
Replacement Wick and Cap
Unit Size:
Each
Product Code. 10251031
Brand: Usbeck Laborgeraete™ 1500
Only null left
Add to basket
Edge
Add to basket
Specifications
Product Type Spirit Burner
Material Stainless Steel
Capacity (Metric) 60 mL
Includes Adjustable Wick and Cap
No. per Pack 1 Ea.
Weight (Metric) 75 g