Learn More
Masterflex™ Stainless Steel Barbed Sanitary Clamp Adapter
Brand: Masterflex™ 30700-60
This item is not returnable.
View return policy
Specifications
Barbed Sanitary Clamp Adapter | |
Stainless Steel | |
Fits 3/4 in. Clamp x 3/8 in. Tubing I.D. | |
1 Pack |