Learn More
Cole Parmer Stainless Steel Beaker
138.67 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
stainless steel beaker
Description
Colel Parmer Stainless Steel Beaker prevents sample contamination
- Constructed of durable 304 medium gauge stainless steel for easy cleaning
- Recessed covers are ideal for maximizing storage space.
- Cover must be purchased separately
Specifications
Capacity (English) | 12 qt. |
Capacity (Metric) | 11.4 L |
Material | Stainless Steel |