Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF750plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ SF750plusB04362
Product Code. 17538365
  • 380.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF750plus Sterilizer
Bottom Drip Tray
Rim: 15 mm
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for SF750plus, 15 mm > For Use With: SF750plus