Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F1
Adjusted to Meet or Exceed OIML F1 Tolerances
35.90 € - 1465.00 €
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16662402
|
Ohaus™
80780307 |
1 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16672402
|
Ohaus™
80780308 |
2 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16682402
|
Ohaus™
80780309 |
5 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16692402
|
Ohaus™
80780310 |
10 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16602412
|
Ohaus™
80780311 |
20 mg |
35.90 €
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
16612412
|
Ohaus™
80780312 |
50 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16622412
|
Ohaus™
80780313 |
100 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16632412
|
Ohaus™
80780314 |
200 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16642412
|
Ohaus™
80780315 |
500 mg |
35.90 €
Each |
In Stock
Log in to see stock. |
|||||
16652412
|
Ohaus™
80780316 |
1 g |
46.15 €
Each |
In Stock
Log in to see stock. |
|||||
16662412
|
Ohaus™
80780317 |
2 g |
46.15 €
Each |
In Stock
Log in to see stock. |
|||||
16672412
|
Ohaus™
80780318 |
5 g |
51.25 €
Each |
In Stock
Log in to see stock. |
|||||
16682412
|
Ohaus™
80780319 |
10 g |
51.25 €
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
16692412
|
Ohaus™
80780320 |
20 g |
61.50 €
Each |
In Stock
Log in to see stock. |
|||||
16602422
|
Ohaus™
80780321 |
50 g |
66.65 €
Each |
In Stock
Log in to see stock. |
|||||
16612422
|
Ohaus™
80780322 |
100 g |
81.80 €
Each |
In Stock
Log in to see stock. |
|||||
16622422
|
Ohaus™
80780323 |
200 g |
103.00 €
Each |
In Stock
Log in to see stock. |
|||||
16632422
|
Ohaus™
80780324 |
500 g |
159.00 €
Each |
In Stock
Log in to see stock. |
|||||
16642422
|
Ohaus™
80780325 |
1 kg |
221.00 €
Each |
In Stock
Log in to see stock. |
|||||
16652422
|
Ohaus™
80780326 |
2 kg |
344.00 €
Each |
In Stock
Log in to see stock. |
|||||
16662422
|
Ohaus™
80780327 |
5 kg |
487.00 €
Each |
In Stock
Log in to see stock. |
|||||
16672422
|
Ohaus™
80780328 |
10 kg |
923.00 €
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
16682422
|
Ohaus™
80780329 |
20 kg |
1465.00 €
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |