Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F2 Tolerances
46.15 € - 1095.00 €
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16682262
|
Ohaus™
30390900 |
1 mg |
46.15 €
Each |
In Stock
Log in to see stock. |
|||||
16672262
|
Ohaus™
30390899 |
2 mg |
46.15 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16662262
|
Ohaus™
30390898 |
5 mg |
46.15 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16652262
|
Ohaus™
30390897 |
10 mg |
46.15 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
16642262
|
Ohaus™
30390896 |
20 mg |
46.15 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
16632262
|
Ohaus™
30390895 |
50 mg |
46.15 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
16622262
|
Ohaus™
30390894 |
100 mg |
46.15 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16612262
|
Ohaus™
30390893 |
200 mg |
46.15 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16602262
|
Ohaus™
30390892 |
500 mg |
46.15 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16692252
|
Ohaus™
30390891 |
1 g |
56.40 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16682252
|
Ohaus™
30390890 |
2 g |
56.40 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16672252
|
Ohaus™
30390889 |
5 g |
61.50 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16662252
|
Ohaus™
30390888 |
10 g |
61.50 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16652252
|
Ohaus™
30390887 |
20 g |
66.65 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16642252
|
Ohaus™
30390886 |
50 g |
71.75 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16632252
|
Ohaus™
30390885 |
100 g |
76.90 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16622252
|
Ohaus™
30390884 |
200 g |
92.25 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16612252
|
Ohaus™
30390883 |
500 g |
123.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
16552592
|
Ohaus™
30390882 |
1 kg |
159.00 €
Each |
In Stock
Log in to see stock. |
|||||
16602252
|
Ohaus™
30390881 |
2 kg |
216.00 €
Each |
In Stock
Log in to see stock. |
|||||
16692242
|
Ohaus™
30390880 |
5 kg |
416.00 €
Each |
In Stock
Log in to see stock. |
|||||
16682242
|
Ohaus™
30390879 |
10 kg |
631.00 €
Each |
In Stock
Log in to see stock. |
|||||
16672242
|
Ohaus™
30390878 |
20 kg |
1095.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |