missing translation for 'onlineSavingsMsg'
Learn More

3M™ Stainless Steel Cleaner & Polish, Aerosol

Product Code. 17158590
Click to view available options
Volume (Metric):
600 mL
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 17158590

Brand: 3M™ 7000042450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for stainless steel, chrome, laminated plastics, and aluminum surfaces. The high gloss, lime-fragranced formula wipes clean with no streaks or buildup, and resists fingerprints.

  • Polishes and protects all stainless steel surfaces in one step
  • Clings to vertical surfaces and leaves a light, non-greasy film that resists fingerprints and markings
  • Water-based with a pleasant citrus fragrance
  • Does not contain harmful chemicals
  • Safe to use in food preparation areas
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleaner/Polish
Form Aerosol
Scent Lime
For Use With (Equipment) Stainless Steel Surfaces
Quantity 12 Pk.
Volume (Metric) 600 mL
compliance-icons
Product Identifier
  • 3M Stainless Steel Cleaner
Signal Word
  • Warning
Hazard Category
  • Flammable and non-flammable aerosols category 2
Hazard Statement
  • H223-Flammable aerosol.
  • H229-Pressurised container: May burst if heated.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Supplemental information
  • MIXTURE LIST-Contains : butane, isobutane, propane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.