missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless Steel Comfort Grinding Jars
875.00 € - 1980.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10052752
|
RETSCH
01.462.0239 |
12 mL | N/A | N/A | N/A | |||||
10515173
|
RETSCH
01.462.0240 |
25 mL |
1000.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
11934372
|
RETSCH
01.462.0321 |
80 mL | N/A | N/A | N/A | |||||
Specifications
Comfort Grinding Jar | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Stainless Steel Comfort Grinding Jars