Learn More
Buerkle™ Stainless Steel Conical QualiRod
Conical hand-drill, conically shaped, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.)
90.05 € - 146.00 €
Specifications
Material Stainless Steel (AISI 304)
Autoclavable Autoclavable
View More Specs
Products 2
Product Code Brand Length (Metric) Blade Price Quantity & Availability  
Product Code Brand Length (Metric) Blade Price Quantity & Availability  
10690543
View Documents
Buerkle™
5311-0001
110 mm
90.05 €
Each
In Stock
to see stock.
Add to basket
13597350
View Documents
Buerkle™
5311-0002
150 mm
146.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
A conically shaped hand-drill, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.).
  • Stainless steel V2A (1.4301)
  • Sterilizable
  • According to EN/ISO 707
Specifications
Specifications
Stainless Steel (AISI 304)
Autoclavable