missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Cylindrical Scoops
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.
23.80 € - 45.05 €
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11553452
|
Fisherbrand™
9010 |
190 mm |
23.80 €
Each |
In Stock
Log in to see stock. |
|||||
11563452
|
Fisherbrand™
9020 |
260 mm |
33.05 €
Each |
In Stock
Log in to see stock. |
|||||
11573452
|
Fisherbrand™
9030 |
500 mL | 325 mm |
31.95 €
Each |
In Stock
Log in to see stock. |
||||
11583452
|
Fisherbrand™
9040 |
1000 mL | 340 mm |
45.05 €
Each |
In Stock
Log in to see stock. |
||||
Description
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.
- Durable, stainless steel, highly polished, and easy to clean cylindrical scoop
- Can be sterilised by autoclaving
- Well-suited for pharmaceutical and environmental sampling
- Ideal for weighing and handling of samples and other materials
- Great accessory to have if you own a weighing scale
- Perfect for weighing anything from liquid to gems and other materials
Specifications
Scoop | |
Stainless Steel | |
Cylindrical |