missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Stainless Steel Disposal Bin
The Stainless Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.
220.00 € - 360.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17886546
|
asecos™
40099 |
20 L |
220.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17896336
|
asecos™
23159 |
35 L |
254.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17836346
|
asecos™
23160 |
50 L |
307.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17896426
|
asecos™
23161 |
65 L |
360.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
- Self-closing lid with damping, large, ergonomically shaped handle on the rear of the container for easy emptying of the container
- Opening by large pedal with anti-slip surface
- FM-certified
Specifications
Gray | |
Stainless Steel | |
Liquid Disposal Can |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title