Learn More
Schneider Gerd™ Stainless Steel Double Micro-spatula
Use this micro spatula for a variety of applications. Schneider Gerd™ Stainless Steel Double Micro-spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 3854
 View more versions of this product
Product Code. 10386741
  • 7.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for working with miniscule amounts of chemicals
  • Two different end shapes, offer increased versatility
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
1 Ea.
Double Ended Micro Spatula
Stainless Steel
Double
5 mm
210 mm