Learn More
BINDER™ Expansion Rack, Includes Calibration
Use this stainless steel expansion rack instead of the standard aluminum expansion rack. Includes calibration.
Brand:  BINDER™ 8612-0011
 View more versions of this product
Product Code. 16616483
  • 631.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel
  • Accessory rack
Specifications
Specifications
Expansion racks - stainless steel expansion rack instead of standard aluminum expansion rack, incl. calibration
Rack
Stainless Steel
BINDER Chambers VD 23, VDL 23
Calibration