Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Holds flasks in place when used with Grant Aqua Pro Shaking Baths
Brand:  Grant Instruments™ SC-100
 View more versions of this product
Product Code. 15365852
  • 77.90 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spring clamp provides security while allowing easy flask insertion/removal
  • For use with OLS and LSB Model Aqua Pro Shaking Water Baths
Specifications
Specifications
100mL Flask
Flask Clamp
Stainless Steel