Learn More
Aalborg™ Stainless Steel Flowmeter Glass Tube
Designed for low flowrates, the model P flowmeter is a precision instrument embodying the inherent simplicity, versatility and economy of the classical variable area meter. Aalborg™ Stainless Steel Flowmeter Glass Tube is particularly suitable for metering carrier gases in chromatography.
Brand:  Aalborg™ PMR1-018121
Product Code. 15350614
  • 756.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of flowtubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
Specifications
Specifications
Flowmeter
13.8 bar
150 mm
121°C
±5%
Stainless Steel
Viton-A™