missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Stainless Steel Forceps

Use Usbeck Laborgeraete™ Stainless Steel Forceps in sterile production environments and cleanrooms. The forceps are designed for repeated sterilization and autoclaving over years of use.

Brand:  Usbeck Laborgeraete™ 3017

Product Code. 10268911

  • 9.11 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tip Style: Straight/blunt point
  • Length: 300mm
  • Weight: 0.1kg
Specifications

Specifications

Straight, Blunt
300 mm
Each
Stainless Steel
Forceps
Forceps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.