Learn More
Nickel Electro™ Stainless Steel Forceps
Straight, blunt round ends. Nickel-Electro™ Stainless Steel Forceps contain serrated jaws.
Brand:  Nickel Electro™ 7117
Product Code. 15699310
  • 12.00 € / 1 pair
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stainless Steel Forceps
  • Straight, serrated jaws
  • Blunt round ends
  • 300mm Length
Specifications
Specifications
Blunt
12 in.
300 mm
300 mm
1 Ea.
Stainless Steel
Laboratory forceps/tweezers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Forceps >