missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Forceps

For use with filter paper. BOCHEM™ Stainless Steel Forceps have 12mm wide points.

Brand:  Bochem™ 1645

Product Code. 11835161

  • 90.75 € / Pack of 10
Estimated Shipment: 16-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with filter paper 
Specifications

Specifications

1 Pack
125 mm
For filter paper
12 mm
Stainless Steel
Forceps
12 mm
Forceps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers