missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Graduated Buckets with Base Ring

Product Code. p-8005456
Click to view available options
Capacity (Metric):
10 L
15 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10137390

Brand: Bochem™ 8315

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Graduated Bucket
Capacity (Metric) 10 L
Material Stainless Steel (18/10)
Diameter (Metric) Outer 280 mm
Height (Metric) 235 mm
Includes With base ring
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.