missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Grinding Balls

Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.

Brand:  RETSCH 224550001

 View more versions of this product

Product Code. 11727664

  • 267.00 € / Pack of 200
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with MM 200 and MM 400 Mixer Mills
Specifications

Specifications

Grinding Balls
4 mm
Reaction Vials
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RETSCH Stainless Steel Grinding Balls > Dia.: 4mm, For Use With: Reaction Vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.