Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
Specifications
Quantity 10/Pk.
Material Stainless Steel
View More Specs
Description
Description
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Specifications
10/Pk.
Stainless Steel