missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Ice Borer

Buerkle™ Stainless Steel Ice Borer is a sampler for deep-frozen and semi-solid substances products in the food industry.

Brand:  Buerkle™ 5323-0200

 View more versions of this product

Product Code. 11866393

  • 439.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilisable
  • 20cm boring depth
  • The sharp sawing crown of the very robust hollow borer rapidly cuts into the sample
  • A cylindrical sample (cross-sectional sample) of up to 20cm can be cut out
Specifications

Specifications

Ice Borer
Autoclavable
Ideal for Deep-Frozen and Semi-Solid Substances Products in The Food Industry
200 mm
Stainless Steel (V4A)
I.D. 16 mm
21 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.