missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Stainless Steel Industrial Scoop
Constructed of high quality stainless steel to make it low cost and easy to use. Sampling Systems™ Stainless Steel Industrial Scoop is ideal for general and pharmaceutical use.
15.35 € - 26.00 €
Product Code | Brand | Capacity (Metric) | Length (Metric) | Length (Metric) Bowl | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Length (Metric) Bowl | Width (Metric) | Price | Quantity & Availability | ||
15839701
|
Sampling Systems™
A643-100 |
100 mL | 180 mm | 115 mm | 60 mm |
15.35 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
15849701
|
Sampling Systems™
A643-200 |
200 mL | 205 mm | 135 mm | 75 mm |
16.85 €
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||
15859701
|
Sampling Systems™
A643-300 |
300 mL | 230 mm | 150 mm | 90 mm |
18.40 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
15869701
|
Sampling Systems™
A643-500 |
500 mL | 270 mm | 180 mm | 110 mm |
26.00 €
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
Description
- Light duty construction
- Stainless Steel
- Fully welded construction
- Easy to clean
Specifications
Scoop | |
Stainless Steel | |
Polished |