Learn More
Socorex™ Stainless Steel Injection Needles
Manufactured with Swiss made quality, Socorex™ Stainless Steel Injection Needles are Luer Lock needles that are reliable and durable. They include a dispensing box for easy and safe removal and dispensing of needles.
Brand:  Socorex™ 371.2050
 View more versions of this product
Product Code. 11807293
  • 62.10 € / Pack of 12
Explore more special offers
Description
Description
  • Stainless steel
  • Sharp bevel
  • Reusable
  • Autoclavable
  • Includes dispensing box with sliding lid for easy and safe needle removal
Specifications
Specifications
Autoclavable
Stainless Steel
Sharp Bevel
2 mm
Injection Needle
14 G
50 mm