missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbolite™ Stainless Steel Inner Chamber Oven
Brand: Carbolite™ LHT5120-230SN+TMP-R38-301
10877.50 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Stainless Steel | |
500°C | |
Digital Temperature Controller | |
2 | |
±0.2°C | |
1030 mm | |
650 mm | |
405 mm | |
208/240 V 3.00 kw |
120 L | |
Oven | |
With 2 Shelves | |
±5°C | |
405 x 480 x 650 mm | |
480 mm | |
670 mm | |
333 lb. | |
208/240 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction