missing translation for 'onlineSavingsMsg'
Learn More

Smeg™ Stainless Steel Instrument Basket

Designed for use with Smeg™ glassware washers. Smeg Stainless Steel Instrument Baskets are suitable for medium-sized instruments.

Brand:  Smeg™ CSK2

 View more versions of this product

Product Code. 13287949

  • 251.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Baskets with stainless steel handles, suitable for medium-sized instruments
  • Mesh openings of 5 x 5mm
  • Dimensions: 225 x 450 x 50mm
Specifications

Specifications

Stainless Steel
Medium-sized instruments
225 x 450 x 50mm
Instrument Basket
Handles
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers