Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
Brand:  Nickel Electro™ 207
 View more versions of this product
Product Code. 11449569
  • 174.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durable stainless steel laboratory beaker
  • 304 spec stainless steel beakers 
  • Will not shatter or chip
  • Suitable for laboratory applications since 
  • All surfaces are non-reacting 
  • Offer zero absorption
Specifications
Specifications
125 mm
Stainless Steel
Spout
2000 mL
180 mm
Beaker