Learn More
Bochem™ Stainless Steel Laboratory Bowl
Bochem™ high form, conical-shaped stainless steel bowls are ideal for use as mixing bowls or for general lab use.
Brand:  Bochem™ 8607
Product Code. 10779155
  • 39.75 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High Form
  • 18/10 Stainless Steel
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
  • Wide range of uses.
Specifications
Specifications
Stainless Steel (18/10)
130 mm
High, Round
5 L
265 mm