Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250μL and larger.
Brand:  Hamilton™ 7806-03
 View more versions of this product
Product Code. 15937335
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Specifications
Specifications
Autoclavable
26 G
0.46 mm
6/Pk.
Removable Syringes (250μL and larger)
Stainless Steel
0.26 mm
Removable Custom Needle
2, 3, or 4