Learn More
Grant Instruments™ Stainless Steel Lid
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
Brand:  Grant Instruments™ LST38
 View more versions of this product
Product Code. 15172625
  • 528.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
  • Stainless steel construction
  • Gabled with plastic handle
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
  • For continuous use with water above 90°C.
Specifications
Specifications
Length: 574
ST38 tank
Stainless Steel