Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7749-04
 View more versions of this product
Product Code. 10443305
  • 210.00 € / Pack of 6
Explore more special offers
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Stainless Steel
1.18 mm
Luer Lock Custom Needle
2, 3, or 4
Standard Tapered
13 G
2.41 mm
9.52 to 304 mm
TLL Syringes and LT or TLL Connectors
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4 > Gauge: 13